Merck
  • Alteration of Gene Expression Profile in Kidney of Spontaneously Hypertensive Rats Treated with Protein Hydrolysate of Blue Mussel (Mytilus edulis) by DNA Microarray Analysis.
Alteration of Gene Expression Profile in Kidney of Spontaneously Hypertensive Rats Treated with Protein Hydrolysate of Blue Mussel (Mytilus edulis) by DNA Microarray Analysis.
PloS one (2015-10-31)
Junli Feng, Zhiyuan Dai, Yanping Zhang, Lu Meng, Jian Ye, Xuting Ma
ZUSAMMENFASSUNG
Marine organisms are rich sources of bioactive components, which are often reported to have antihypertensive effects. However, the underlying mechanisms have yet to be fully identified. The aim of this study was to investigate the antihypertensive effect of enzymatic hydrolysis of blue mussel protein (HBMP) in rats. Peptides with in vitro ACE inhibitory activity were purified from HBMP by ultrafiltration, gel filtration chromatography and reversed-phase high performance liquid chromatography. And the amino acid sequences of isolated peptides were estimated to be Val-Trp, Leu-Gly-Trp, and Met-Val-Trp-Thr. To study its in vivo action, spontaneously hypertensive rats (SHRs) were orally administration with high- or low-dose of HBMP for 28 days. Major components of the renin-angiotensin (RAS) system in serum of SHRs from different groups were analyzed, and gene expression profiling were performed in the kidney of SHRs, using the Whole Rat Genome Oligonucleotide Microarray. Results indicated although genes involved in RAS system were not significantly altered, those related to blood coagulation system, cytokine and growth factor, and fatty acids metabolism were remarkablely changed. Several genes which were seldom reported to be implicated in pathogenesis of hypertension also showed significant expression alterations after oral administration of HBMP. These data provided valuable information for our understanding of the molecular mechanisms that underlie the potential antihypertensive activities of HBMP, and will contribute towards increased value-added utilization of blue mussel protein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Angiotensin-konvertierendes Enzym aus Kaninchenlunge, ≥2.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Hippursäure, 98%
Sigma-Aldrich
N-Hippuryl-His-Leu Hydrat, powder, ≥98% (HPLC)
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
N-Hexanoyl-L-homoserin-lacton, ≥96% (HPLC)
Sigma-Aldrich
Schwefelsäure, puriss., meets analytical specification of Ph. Eur., BP, 95-97%