Merck
  • Ciliary and centrosomal defects associated with mutation and depletion of the Meckel syndrome genes MKS1 and MKS3.
Ciliary and centrosomal defects associated with mutation and depletion of the Meckel syndrome genes MKS1 and MKS3.
Human molecular genetics (2009-06-12)
Rachaneekorn Tammachote, Cynthia J Hommerding, Rachel M Sinders, Caroline A Miller, Peter G Czarnecki, Amanda C Leightner, Jeffrey L Salisbury, Christopher J Ward, Vicente E Torres, Vincent H Gattone, Peter C Harris
ZUSAMMENFASSUNG
Meckel syndrome (MKS) is a lethal disorder characterized by renal cystic dysplasia, encephalocele, polydactyly and biliary dysgenesis. It is highly genetically heterogeneous with nine different genes implicated in this disorder. MKS is thought to be a ciliopathy because of the range of phenotypes and localization of some of the implicated proteins. However, limited data are available about the phenotypes associated with MKS1 and MKS3, and the published ciliary data are conflicting. Analysis of the wpk rat model of MKS3 revealed functional defects of the connecting cilium in the eye that resulted in lack of formation of the outer segment, whereas infertile wpk males developed spermatids with very short flagella that did not extend beyond the cell body. In wpk renal collecting duct cysts, cilia were generally longer than normal, with additional evidence of cells with multiple primary cilia and centrosome over-duplication. Kidney tissue and cells from MKS1 and MKS3 patients showed defects in centrosome and cilia number, including multi-ciliated respiratory-like epithelia, and longer cilia. Stable shRNA knockdown of Mks1 and Mks3 in IMCD3 cells induced multi-ciliated and multi-centrosomal phenotypes. These studies demonstrate that MKS1 and MKS3 are ciliopathies, with new cilia-related eye and sperm phenotypes defined. MKS1 and MKS3 functions are required for ciliary structure and function, including a role in regulating length and appropriate number through modulating centrosome duplication.