Merck
  • Surgical cytoreduction restores the antitumor efficacy of a Listeria monocytogenes vaccine in malignant pleural mesothelioma.
Surgical cytoreduction restores the antitumor efficacy of a Listeria monocytogenes vaccine in malignant pleural mesothelioma.
Immunology letters (2015-05-23)
Gregory T Kennedy, Brendan F Judy, Pratik Bhojnagarwala, Edmund K Moon, Zvi G Fridlender, Steven M Albelda, Sunil Singhal
ZUSAMMENFASSUNG
Recent studies suggest that immunotherapy may offer a promising treatment strategy for early-stage malignant pleural mesothelioma (MPM), but advanced tumor burden may limit the efficacy of immunotherapy. Therefore, we hypothesized that surgical cytoreduction could restore the efficacy of vaccine-based immunotherapy for MPM. We developed a murine model of MPM through transduction of a mesothelioma cell line with mesothelin. We used this model to evaluate the efficacy of a Listeria monocytogenes vaccine expressing mesothelin. Tumor growth was significantly inhibited at four weeks in animals vaccinated two weeks prior to tumor cell inoculation as compared to those given an empty vector control (1371 ± 420 mm(3) versus 405 ± 139 mm(3); p < 0.01). Mice vaccinated one week prior to tumor challenge also displayed significant reduction in tumor volume (1227 ± 406 mm(3) versus 309 ± 173 mm(3); p < 0.01). The vaccine had no effect when administered concurrently with tumor challenge, or after tumors were established. Flow cytometry showed reduced mesothelin expression in large tumors, as well as tumor-associated immunosuppression due to increased myeloid derived suppressor cells (MDSCs). These factors may have limited vaccine efficacy for advanced disease. Surgical cytoreduction of established tumors restored the antitumor potency of the therapeutic vaccine, with significantly reduced tumor burden at post-operative day 18 (397 ± 103 mm(3) versus 1047 ± 258 mm(3); p < 0.01). We found that surgery reduced MDSCs to levels comparable to those in tumor-naïve mice. This study demonstrates that cytoreduction surgery restores the efficacy of cancer vaccines for MPM by reducing tumor-related immunosuppression that impairs immunotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining