Merck
Breakdowns of eye movement control toward smoking cues in young adult light smokers.
Addictive behaviors (2015-09-26)
Gregory J DiGirolamo, Ellen J Sophis, Jennifer L Daffron, Gerardo Gonzalez, Mauricio Romero-Gonzalez, Sean A Gillespie
ZUSAMMENFASSUNG
Many studies suggest that dependent smokers have a preference or attentional bias toward smoking cues. The purpose of this study was to test the ability of infrequent non-dependent light smokers to control their eye movements by look away from smoking cues. Poor control in the lightest of smokers would suggest nicotine cue-elicited behavior occurring even prior to nicotine dependency as measured by daily smoking. 17 infrequent non-dependent light smokers and 17 lifetime non-smokers performed an antisaccade task (look away from suddenly appearing cue) on smoking, alcohol, neutral, and dot cues. The light smokers, who were confirmed light smokers and non-dependent (MFaegerström Dependency Score=0.35), were significantly worse at controlling their eye movements to smoking cues relative to both neutral cues (p<.04) and alcohol cues (p<.02). Light smokers made significantly more errors to smoking cues than non-smokers (p<.004). These data suggest that prior to developing clinical symptoms of severe dependence or progressing to heavier smoking (e.g., daily smoking), the lightest of smokers are showing a specific deficit in control of nicotine cue-elicited behavior.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N′-Methylenbisacrylamid, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenbis(acrylamid), 99%
Sigma-Aldrich
N,N′-Methylenbisacrylamid -Lösung, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
N,N′-Methylenbisacrylamid, suitable for electrophoresis (after filtration or allowing insolubles to settle)