Merck
  • Effect of Quercetin on Cell Cycle and Cyclin Expression in Ovarian Carcinoma and Osteosarcoma Cell Lines.
Effect of Quercetin on Cell Cycle and Cyclin Expression in Ovarian Carcinoma and Osteosarcoma Cell Lines.
Natural product communications (2015-10-06)
Daniela Catanzaro, Eugenio Ragazzi, Caterina Vianello, Laura Caparrotta, Monica Montopoli
ZUSAMMENFASSUNG
Resistance to chemotherapeutic drugs is a major problem in cancer treatment. The search for new interventions able to overcome this resistance may involve compounds of natural origin, such as flavonoids, ubiquitously present in many foods. In the present study, the cytotoxic effects and cell cycle modulation of the flavonoid quercetin were investigated in ovarian carcinoma (SKOV3) and osteosarcoma (U2OS) human cell lines and in their cisplatin (CDDP)-resistant counterparts (SKOV3/CDDP and U2OSPt cells, respectively). Quercetin (10-50 μM) caused evident changes in the distribution of cell cycle phases in the CDDP-resistant SKOV3/CDDP ovarian cell line. The levels of cyclin D1 and cyclin B1 were determined by means of Western blot in all cell lines incubated with quercetin (50 μM) for 48 hours. The cyclin D1 expression was significantly decreased following the treatment with quercetin in SKOV3 and U2OSPt cells, but not in SKOV3/CDDP and U2OS cells. The reduction of cyclin D1 level could be linked to the G1/S phase alteration found in quercetin-treated cells. Although cyclin B1 is required for G2/M phase, and despite our observation that quercetin influenced the G2/M phase of cell cycle, the flavonoid did not affect cyclin B1 levels in all cell lines, indicating the involvement of other possible mechanisms. These results suggest that quercetin, exceeding the resistance to CDDP, might become an interesting tool to evaluate cytotoxic activity in combination with chemotherapy drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Propidiumjodid -Lösung