Merck
Mmu-miR-126a-3p plays a role in murine embryo implantation by regulating Itga11.
Reproductive biomedicine online (2015-07-22)
Zhengyu Li, Jia Jia, Jinhai Gou, Aiping Tong, Xinyu Liu, Xia Zhao, Tao Yi
ZUSAMMENFASSUNG
miR-126a-3p has been found to be specifically up-regulated in the process of murine embryo implantation. This study aimed to further clarify the role of miR-126a-3p in embryo implantation. The expression of miR-126a-3p in implantation sites was significantly higher than that in interimplantation sites (P = 0.009). Its expression dynamics in a series of models, including pseudopregnancy, delayed implantation and artificial decidualization, suggested that the induction of miR-126a-3p is dependent on hormonal signalling and decidualization of the endometrium. Bioinformatic analysis predicted and luciferase activity assay confirmed that Itga11, a member of the integrin family, was a target gene of miR-126a-3p. miR-126a-3p bound to the 3' untranslated region of Itga11 and regulated Itga11 by inhibiting mRNA translation and affecting mRNA stability. Transwell assay showed that miR-126a-3p promoted cell migratory and invasive capacity in vitro. Loss of function of miR-126a-3p significantly reduced the number of implantation sites in vivo (P = 0.013). Collectively, miR-126a-3p may play a major role in embryo implantation by regulating Itga11, possibly through impairing cell migratory and invasive capacity. These findings should contribute to a better understanding of the miRNA-based mechanism of embryo implantation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture