Merck
  • Minimal sample requirement for highly multiplexed protein quantification in cell lines and tissues by PCT-SWATH mass spectrometry.
Minimal sample requirement for highly multiplexed protein quantification in cell lines and tissues by PCT-SWATH mass spectrometry.
Proteomics (2015-08-20)
Shiying Shao, Tiannan Guo, Chiek Ching Koh, Silke Gillessen, Markus Joerger, Wolfram Jochum, Ruedi Aebersold
ZUSAMMENFASSUNG
The amount of sample available for clinical and biological proteomic research is often limited and thus significantly restricts clinical and translational research. Recently, we have integrated pressure cycling technology (PCT) assisted sample preparation and SWATH-MS to perform reproducible proteomic quantification of biopsy-level tissue samples. Here, we further evaluated the minimal sample requirement of the PCT-SWATH method using various types of samples, including cultured cells (HeLa, K562, and U251, 500 000 to 50 000 cells) and tissue samples (mouse liver, heart, brain, and human kidney, 3-0.2 mg). The data show that as few as 50 000 human cells and 0.2-0.5 mg of wet mouse and human tissues produced peptide samples sufficient for multiple SWATH-MS analyses at optimal sample load applied to the system. Generally, the reproducibility of the method increased with decreasing tissue sample amounts. The SWATH maps acquired from peptides derived from samples of varying sizes were essentially identical based on the number, type, and quantity of identified peptides. In conclusion, we determined the minimal sample required for optimal PCT-SWATH analyses, and found smaller sample size achieved higher quantitative accuracy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C