Merck
  • ILM peeling technique influences the degree of a dissociated optic nerve fibre layer appearance after macular hole surgery.
ILM peeling technique influences the degree of a dissociated optic nerve fibre layer appearance after macular hole surgery.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2014-07-17)
David H W Steel, Christiana Dinah, Maged Habib, Kathryn White
ZUSAMMENFASSUNG
We sought to assess the effect of two different internal limiting membrane [ILM] peeling techniques carried out during surgery for idiopathic macular holes on the postoperative extent of a dissociated optic nerve fibre layer appearance [DONFL]. We collected prospective data of surgical records, videos, and pre- and postoperative imaging of a consecutive series of patients undergoing surgery for idiopathic macular hole with one of two surgeons. One surgeon used a forceps pinch-peel technique to peel the ILM, whereas the other surgeon used a diamond dusted membrane scraper. The extent of any DONFL was measured using spectral domain optical coherence tomography and blue reflectance imaging at three months postoperatively. A proportion of the ILMs removed were examined with transmission electron microscopy. Fifty-seven patients were studied, with 41 in the forceps group and 16 in the scraper group. The groups were well matched, with no significant difference in any preoperative parameters. Some degree of DONFL was observed on the 3-month blue reflectance images in 88 % of the forceps group and 100 % of the scraper group [p = 0.14]. There was a significant difference in the total number of depressions in the nerve fibre layer typical of DONFL on OCT between the two groups [p = 0.001], and general regression analysis showed that the peeling technique used had the only significant association with the degree of DONFL observed. Electron microscopy showed large patches of cellular debris on the retinal side of the peeled ILM in 3 out of 4 cases in the scraper group and 1 out of 12 cases in the forceps group. ILM peeling technique and possibly other surgeon-specific factors appear to influence the extent of DONFL observed after ILM peeling macular hole surgery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications