Merck
GC-MS based metabolomics study of stems and roots of Ephedra sinica.
Journal of pharmaceutical and biomedical analysis (2015-05-26)
Mengying Lv, Jianbo Sun, Min Wang, Wanqiu Huang, Hongyan Fan, Fengguo Xu, Zunjian Zhang
ZUSAMMENFASSUNG
Therapeutic effects of herbal medicines differ greatly due to the use of different anatomical parts or processing methods in traditional Chinese medicine, and Ephedra sinica (ES) is just a case in point. To better understand different traditional uses of the stems (known as Mahuang, MH) and roots (known as Mahuanggen, MHG) of ES, their therapeutic material basis should be investigated. In this study, ephedrine alkaloids were profiled simultaneously with primary metabolites using GC-MS based metabolomics. Ephedrine (E) has been reported to be the major bioactive constituent in MH for the treatment of asthma. The results showed that compared with MH, MHG contained much lower levels of five ephedrine alkaloids, which may well explain that MHG has not been used as an antiasthmatic. Additionally, these pharmacologically important ephedrine alkaloids exhibited strong positive correlation with five primary metabolites. In conclusion, this study facilitates better understanding of different traditional uses of MH and MHG.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäureanhydrid, ACS reagent, ≥98.0%
Sigma-Aldrich
Essigsäureanhydrid, 99.5%
Sigma-Aldrich
Pyridin, ≥99%
Supelco
N-Methyl-bis-trifluoracetamid, derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Trifluoracetamid, 97%
Sigma-Aldrich
Essigsäureanhydrid, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.