Merck
  • Fluorescence Enhancement of Nanoraspberry Hot-spot Source Composed of Gold Nanoparticles and Aniline Oligomers.
Fluorescence Enhancement of Nanoraspberry Hot-spot Source Composed of Gold Nanoparticles and Aniline Oligomers.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-06-13)
Takamasa Kinoshita, Dung Quang Nguyen, Tomoaki Nishino, Hidenobu Nakao, Hiroshi Shiigi, Tsutomu Nagaoka
ZUSAMMENFASSUNG
In this study, we examined raspberry-shaped organic/inorganic hybrid structure for potential development of a nanoantenna system capable of detecting and labeling biomolecules. The structure is characterized by a high density of gold nanoparticles (AuNPs) separated by closely packed aniline oligomers that serve as a linkage between adjacent particles. In particular, the structure was based on repeated sequences of AuNP-aniline oligomer-AuNP in a three-dimensional arrangement, which enabled the creation of optical hot spots that can hold multiple molecules. We examine the expression of such features by focusing on the structure and characteristics of the hybrid. We demonstrate that these optical hot spots enhance the dye fluorescence without quenching. As a result, we were able to create a nanoantenna structure enabling the efficient use of light.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Sigma-Aldrich
Anilin, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumborhydrid, granular, 99.99% trace metals basis
Sigma-Aldrich
Natriumborhydrid, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Kaliumcarbonat, 99.995% trace metals basis
Sigma-Aldrich
Kaliumcarbonat, powder, 99.99% trace metals basis
Sigma-Aldrich
Anilin, ReagentPlus®, 99%
Sigma-Aldrich
Natriumborhydrid, granular, 10-40 mesh, 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Natriumborhydrid, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Kaliumcarbonat, meets USP testing specifications
Sigma-Aldrich
Kaliumcarbonat, BioUltra, anhydrous, ≥99.0% (T)