Merck
Aligned Carbon Nanotube Thin Films from Liquid Crystal Polyelectrolyte Inks.
ACS applied materials & interfaces (2015-10-30)
Daniel D Tune, Adam J Blanch, Cameron J Shearer, Katherine E Moore, Moritz Pfohl, Joseph G Shapter, Benjamin S Flavel
ZUSAMMENFASSUNG
Single walled carbon nanotube thin films are fabricated by solution shearing from high concentration sodium nanotubide polyelectrolyte inks. The solutions are produced by simple stirring of the nanotubes with elemental sodium in dimethylacetamide, and the nanotubes are thus not subject to any sonication-induced damage. At such elevated concentrations (∼4 mg mL(-1)), the solutions exist in the liquid crystal phase and during deposition this order is transferred to the films, which are well aligned in the direction of shear with a 2D nematic order parameter of ∼0.7 determined by polarized absorption measurements. Compared to similarly formed films made from superacids, the polyelectrolyte films contain smaller bundles and a much narrower distribution of bundle diameters. After p-doping with an organic oxidizer, the films exhibit a very high DC electrical to optical conductivity ratio of σ(DC)/σ(OP) ∼ 35, corresponding to a calculated DC conductivity of over 7000 S cm(-1). When very thin (T550 ∼ 96%), smooth (RMS roughness, R(q) ∼ 2.2 nm), and highly aligned films made via this new route are used as the front electrodes of carbon nanotube-silicon solar cells, the power conversion efficiency is almost an order of magnitude greater than that obtained when using the much rougher (R(q) ∼ 20-30 nm) and less conductive (peak σ(DC)/σ(OP) ∼ 2.5) films formed by common vacuum filtration of the same starting material, and having the same transmittance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Argon-40Ar, 99.95 atom %