Merck
  • Polydopamine meets solid-state nanopores: a bioinspired integrative surface chemistry approach to tailor the functional properties of nanofluidic diodes.
Polydopamine meets solid-state nanopores: a bioinspired integrative surface chemistry approach to tailor the functional properties of nanofluidic diodes.
Journal of the American Chemical Society (2015-04-17)
Gonzalo Pérez-Mitta, Jimena S Tuninetti, Wolfgang Knoll, Christina Trautmann, María Eugenia Toimil-Molares, Omar Azzaroni
ZUSAMMENFASSUNG
The ability to modulate the surface chemical characteristics of solid-state nanopores is of great interest as it provides the means to control the macroscopic response of nanofluidic devices. For instance, controlling surface charge and polarity of the pore walls is one of the most important applications of surface modification that is very relevant to attain accurate control over the transport of ions through the nanofluidic architecture. In this work, we describe a new integrative chemical approach to fabricate nanofluidic diodes based on the self-polymerization of dopamine (PDOPA) on asymmetric track-etched nanopores. Our results demonstrate that PDOPA coating is not only a simple and effective method to modify the inner surface of polymer nanopores fully compatible with the fabrication of nanofluidic devices but also a versatile platform for further integration of more complex molecules through different covalent chemistries and self-assembly processes. We adjusted the chemical modification strategy to obtain various configurations of the pore surface: (i) PDOPA layer was used as primer, precursor, or even responsive functional coating; (ii) PDOPA layer was used as a platform for anchoring chemical functions via the Michael addition reaction; and (iii) PDOPA was used as a reactive layer inducing the metallization of the pore walls through the in situ reduction of metallic precursors present in solution. We believe that the transversal concept of integrative surface chemistry offered by polydopamine in combination with the remarkable physical characteristics of asymmetric nanopores constitutes a new framework to design multifunctional nanofluidic devices employing soft chemistry-based nanofunctionalization techniques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
(+)-Abscisinsäure, ≥98% (HPLC)