Merck
  • Characterization of efflux transport proteins of the human choroid plexus papilloma cell line HIBCPP, a functional in vitro model of the blood-cerebrospinal fluid barrier.
Characterization of efflux transport proteins of the human choroid plexus papilloma cell line HIBCPP, a functional in vitro model of the blood-cerebrospinal fluid barrier.
Pharmaceutical research (2015-05-20)
Alexandra Bernd, Melanie Ott, Hiroshi Ishikawa, Horst Schroten, Christian Schwerk, Gert Fricker
ZUSAMMENFASSUNG
To characterize the human choroid plexus (CP) papilloma cell line HIBCPP with respect to ABC export protein expression and function in order to evaluate its use as an in vitro model to study carrier-mediated transport processes at the CP. Expression profiles of ABC transporters were studied by quantitative real-time PCR and Western Blot analysis. Functionality of transporters was investigated by means of uptake experiments and permeation studies carried out on permeable filter systems. In addition, immunohistochemistry served to study localization of ABCC1 and ABCC4. Both qPCR and Western Blot revealed that ABC transporters known to be expressed in CP are also expressed in HIBCPP cells. Immunohistochemistry confirmed basolateral expression of ABCC1. Functionality of ABCC1, ABCC4, ABCB1 and ABCG2 could be shown in uptake assays. Altogether, the HIBCPP cells promise to be a functional and relevant in vitro tool to investigate transport processes at the blood-cerebrospinal fluid barrier.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Rhodamin 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Rhodamin 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Sulforhodamin 101, Dye content ~95 %
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)