Merck
  • Inhibition of tumor progression during allergic airway inflammation in a murine model: significant role of TGF-β.
Inhibition of tumor progression during allergic airway inflammation in a murine model: significant role of TGF-β.
Cancer immunology, immunotherapy : CII (2015-06-17)
Belen Tirado-Rodriguez, Guillermina Baay-Guzman, Rogelio Hernandez-Pando, Gabriela Antonio-Andres, Mario I Vega, Leticia Rocha-Zavaleta, Laura C Bonifaz, Sara Huerta-Yepez
ZUSAMMENFASSUNG
TGF-β is an important mediator of pulmonary allergic inflammation, and it has been recently reported to be a potential inhibitor of lung tumor progression. The correlation between cancer and allergic inflammatory diseases remains controversial. Thus, the aim of the present study was to evaluate the effects of pulmonary allergic inflammation and in particular the role of TGF-β on cancer progression. Cancer cells were implanted in a BALB/c mice model of allergic airway inflammation, and tumor growth was measured. Apoptosis was evaluated by TUNEL assay, and TGF-β was measured by ELISA. Expression of proliferating cell nuclear antigen, TGF-β, TGF-β receptors I and II, phospho-Smad2 and phospho-Smad4 was evaluated by immunohistochemistry and quantified using digital pathology. The effect of a TGF-β activity inhibitor and recombinant TGF-β on tumor growth was analyzed. The effect of exogenous TGF-β on cell proliferation and apoptosis was evaluated in vitro. Mice with allergic airway inflammation exhibited decreased tumor volumes due to cell proliferation inhibition and increased apoptosis. TGF-β was increased in the sera and tumor tissues of allergic mice. TGF-β activity inhibition increased tumor progression in allergic mice by enhancing proliferation and decreasing apoptosis of tumor cells. The administration of TGF-β resulted in reduced tumor growth. This study is the first to establish an inverse relationship between allergic airway inflammation and tumor progression. This effect appears to be mediated by TGF-β, which is overexpressed in tumor cells during pulmonary allergic inflammation. This study indicates that TGF-β is a potential target for antitumor therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Benzamid, 99%
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazol, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioXtra, ≥98.0%
Sigma-Aldrich
Benzamid, purified by sublimation, ≥99.5%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri