Merck
  • Tissue and sex specificities in Ca2+ handling by isolated mitochondria in conditions avoiding the permeability transition.
Tissue and sex specificities in Ca2+ handling by isolated mitochondria in conditions avoiding the permeability transition.
Experimental physiology (2015-06-23)
Hanan Chweih, Roger F Castilho, Tiago R Figueira
ZUSAMMENFASSUNG
What is the central question of this study? The assessment of Ca(2+) handling by isolated mitochondria can be biased by dysfunctions secondary to Ca(2+) -induced mitochondrial permeability transition (MPT). As a result of this uncertainty and the differing experimental conditions between studies, the tissue and sex diversities in mitochondrial Ca(2+) transport are still unsettled questions. What is the main finding and its importance? If MPT is not prevented during Ca(2+) transport assays, some measured variables are biased. Accounting for the implied importance of preventing MPT, we observed substantial tissue specificities in the mitochondrial Ca(2+) handling, particularly in the Ca(2+) efflux pathways. The characteristics of mitochondria, including their Ca(2+) transport functions, may exhibit tissue specificity and sexual dimorphism. Given that measurements of Ca(2+) handling by isolated mitochondria may be biased by dysfunction secondary to Ca(2+) -induced mitochondrial permeability transition (MPT) pore opening, this study evaluated the extent to which MPT inhibition by ciclosporin affected the measurement of Ca(2+) transport in isolated rat liver mitochondria. The results indicate that the steady-state levels of external Ca(2+) and the rates of mitochondrial Ca(2+) efflux through the selective pathways can be overestimated by up to fourfold if MPT pore opening is not prevented. We analysed Ca(2+) transport in isolated mitochondria from the liver, skeletal muscle, heart and brain of male and female rats in incubation conditions containing MPT inhibitors, NAD-linked substrates and relevant levels of free Ca(2+), Mg(2+) and Na(+). The Ca(2+) influx rates were similar among the samples, except that the liver mitochondria displayed values fourfold higher. In contrast, the Ca(2+) efflux rates exhibited more tissue diversity, especially in the presence of Na(+). Interestingly, the Na(+)-independent Ca(2+) efflux was highest in the heart mitochondria (∼ 4 nmol mg(-1) min(-1)), thus challenging the view that cardiac mitochondrial Ca(2+) efflux relies almost exclusively on a Na(+)-dependent pathway. Sex specificity was observed in only two kinetic indexes of heart mitochondrial Ca(2+) homeostasis and in the ADP-stimulated respiration of liver mitochondria (∼ 20% higher in females). The present study shows the methodological importance of preventing MPT when measuring the properties and the physiological variability of the Ca(2+) handling by isolated mitochondria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Ruthenium auf Kohlenstoff, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Adenin, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Rutheniumschwarz
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, bar, 25mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%