Merck
  • Development and Validation of an HPLC-UV Method for Determination of Eight Phenolic Compounds in Date Palms.
Development and Validation of an HPLC-UV Method for Determination of Eight Phenolic Compounds in Date Palms.
Journal of AOAC International (2015-11-04)
Fuad Al-Rimawi, Imad Odeh
ZUSAMMENFASSUNG
A simple, precise, accurate, and selective method was developed and validated for determination of eight phenolic compounds (gallic acid, p-hydroxybenzoic acid, vanilic acid, caffeic acid, syringic acid, p-coumaric acid, ferulic acid, and sinapic acid) in date palms. Separation was achieved on an RP C18 column using the mobile phase methanol-water with 2% acetic acid (18+82, v/v). This method was validated according to the requirements for new methods, which include accuracy, precision, selectivity, robustness, LOD, LOQ, linearity, and range. The method demonstrated good linearity over the range 1-1000 ppm of gallic acid, p-hydroxybenzoic acid, vanilic acid, caffeic acid, and syringic acid with r2 greater than 0.99, and in the range of 3-1000 ppm for p-coumaric acid, ferulic acid, and sinapic acid with r2 greater than 0.99. The recovery of the eight phenolic compounds ranged from 97.1 to 102.2%. The method is selective because adjacent peaks of phenolic compounds were well separated with good resolution. The degree of reproducibility of the results obtained as a result of small deliberate variations in the method parameters and by changing analytical operators proved that the method is robust and rugged.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Syringasäure, ≥95% (HPLC)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C