Merck
  • Responsiveness of G protein-coupled odorant receptors is partially attributed to the activation mechanism.
Responsiveness of G protein-coupled odorant receptors is partially attributed to the activation mechanism.
Proceedings of the National Academy of Sciences of the United States of America (2015-12-03)
Yiqun Yu, Claire A de March, Mengjue J Ni, Kaylin A Adipietro, Jérôme Golebiowski, Hiroaki Matsunami, Minghong Ma
ZUSAMMENFASSUNG
Mammals detect and discriminate numerous odors via a large family of G protein-coupled odorant receptors (ORs). However, little is known about the molecular and structural basis underlying OR response properties. Using site-directed mutagenesis and computational modeling, we studied ORs sharing high sequence homology but with different response properties. When tested in heterologous cells by diverse odorants, MOR256-3 responded broadly to many odorants, whereas MOR256-8 responded weakly to a few odorants. Out of 36 mutant MOR256-3 ORs, the majority altered the responses to different odorants in a similar manner and the overall response of an OR was positively correlated with its basal activity, an indication of ligand-independent receptor activation. Strikingly, a single mutation in MOR256-8 was sufficient to confer both high basal activity and broad responsiveness to this receptor. These results suggest that broad responsiveness of an OR is at least partially attributed to its activation likelihood.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder