Merck
Acylated flavonol diglucosides from Ammania auriculata.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2015-04-10)
Mahmoud Nawwar, Nahla Ayoub, Mohamed El-Raey, Soumaya Zaghloul, Amani Hashem, Eman Mostafa, Omayma Eldahshan, Ulrike Lindequist, Michael W Linscheid
ZUSAMMENFASSUNG
Chemical investigation of the extract of the whole Ammania auriculata plant resulted in the identification of 13 polyphenols, including the hitherto unknown flavonoids, kaempferol 3-O-β-(6″-galloylglucopyranoside)-7-O-β-glucopyranoside, and its quercetin analogue. The structures of all isolates were elucidated by conventional methods, spectroscopic analysis, including 1D and 2D NMR, and by HRESI-MS as well.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder