Merck
  • Enhanced Field Emission from a Carbon Nanotube Array Coated with a Hexagonal Boron Nitride Thin Film.
Enhanced Field Emission from a Carbon Nanotube Array Coated with a Hexagonal Boron Nitride Thin Film.
Small (Weinheim an der Bergstrasse, Germany) (2015-04-29)
Xiaoxia Yang, Zhenjun Li, Feng He, Mingju Liu, Bing Bai, Wei Liu, Xiaohui Qiu, Hang Zhou, Chi Li, Qing Dai
ZUSAMMENFASSUNG
A high-quality field emission electron source made of a highly ordered array of carbon nanotubes (CNTs) coated with a thin film of hexagonal boron nitride (h-BN) is fabricated using a simple and scalable method. This method offers the benefit of reproducibility, as well as the simplicity, safety, and low cost inherent in using B(2)O(3) as the boron precursor. Results measured using h-BN-coated CNT arrays are compared with uncoated control arrays. The optimal thickness of the h-BN film is found to be 3 nm. As a result of the incorporation of h-BN, the turn-on field is found to decrease from 4.11 to 1.36 V μm(-1), which can be explained by the significantly lower emission barrier that is achieved due to the negative electron affinity of h-BN. Meanwhile, the total emission current is observed to increase from 1.6 to 3.7 mA, due to a mechanism that limits the self-current of any individual emitting tip. This phenomenon also leads to improved emission stability and uniformity. In addition, the lifetime of the arrays is improved as well. The h-BN-coated CNT array-based field emitters proposed in this work may open new paths for the development of future high-performance vacuum electronic devices.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bor, ≥95% (boron), amorphous powder
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Bor, crystalline, 1 cm, 99.7% trace metals basis
Bor, monofilament, 10m, diameter 0.2mm
Bor, monofilament, 5m, diameter 0.2mm
Bor, monofilament, 10m, diameter 0.1mm
Bor, monofilament, 200m, diameter 0.1mm
Bor, monofilament, 20m, diameter 0.2mm
Bor, monofilament, 50m, diameter 0.2mm
Bor, monofilament, 100m, diameter 0.1mm
Bor, monofilament, 50m, diameter 0.1mm
Bor, monofilament, 5m, diameter 0.1mm
Bor, monofilament, 20m, diameter 0.1mm