Merck
  • The effect of bisphosphonate treatment on the biochemical and cellular events during bone remodelling in response to microinjury stimulation.
The effect of bisphosphonate treatment on the biochemical and cellular events during bone remodelling in response to microinjury stimulation.
European cells & materials (2015-11-29)
L E Mulcahy, C M Curtin, R J McCoy, F J O'Brien, D Taylor, T C Lee, G P Duffy
ZUSAMMENFASSUNG
Osteoporosis is one of the most prevalent bone diseases worldwide and is characterised by high levels of bone turnover, a marked loss in bone mass and accumulation of microdamage, which leads to an increased fracture incidence that places a huge burden on global health care systems. Bisphosphonates have been used to treat osteoporosis and have shown great success in conserving bone mass and reducing fracture incidence. In spite of the existing knowledge of the in vivo responses of bone to bisphosphonates, the cellular responses to these drugs have yet to be fully elucidated. In vitro model systems that allow the decoupling of complex highly integrated events, such as bone remodelling, provide a tool whereby these biological processes may be studied in a more simplified context. This study firstly utilised an in vitro model system of bone remodelling and comprising all three major cell types of the bone (osteocytes, osteoclasts and osteoblasts), which was representative of the bone's capacity to sense microdamage and subsequently initiate a basic multicellular unit response. Secondly, this system was used to study the effect of two commonly utilised aminobisphosphonate treatments for osteoporosis, alendronate and zoledronate. We demonstrated that microinjury to osteocyte networks being treated with bisphosphonates modulates receptor activator of nuclear factor kappa-B ligand and osteoprotegerin activity, and subsequently osteoclastogenesis. Furthermore, bisphosphonates increased the osteogenic potential following microinjury. Thus, we have shown for the first time that bisphosphonates act at all three stages of bone remodelling, from microinjury to osteoclastogenesis and ultimately osteogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
SAFC
L-Glutamin
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis