Merck
  • Analysis of DDT and its metabolites in soil and water samples obtained in the vicinity of a closed-down factory in Bangladesh using various extraction methods.
Analysis of DDT and its metabolites in soil and water samples obtained in the vicinity of a closed-down factory in Bangladesh using various extraction methods.
Environmental monitoring and assessment (2015-11-13)
M N U Al Mahmud, Farzana Khalil, Md Musfiqur Rahman, M I R Mamun, Mohammad Shoeb, A M Abd El-Aty, Jong-Hyouk Park, Ho-Chul Shin, Nilufar Nahar, Jae-Han Shim
ZUSAMMENFASSUNG
This study was conducted to monitor the spread of dichlorodiphenyltrichloroethane (DDT) and its metabolites (dichlorodiphenyldichloroethylene (DDE), dichlorodiphenyldichloroethane (DDD)) in soil and water to regions surrounding a closed DDT factory in Bangladesh. This fulfillment was accomplished using inter-method and inter-laboratory validation studies. DDTs (DDT and its metabolites) from soil samples were extracted using microwave-assisted extraction (MAE), supercritical fluid extraction (SFE), and solvent extraction (SE). Inter-laboratory calibration was assessed by SE, and all methods were validated by intra- and inter-day accuracy (expressed as recovery %) and precision (expressed as relative standard deviation (RSD)) in the same laboratory, at three fortified concentrations (n = 4). DDTs extracted from water samples by liquid-liquid partitioning and all samples were analyzed by gas chromatography (GC)-electron capture detector (ECD) and confirmed by GC/mass spectrometry (GC/MS). Linearities expressed as determination coefficients (R (2)) were ≥0.995 for matrix-matched calibrations. The recovery rate was in the range of 72-120 and 83-110%, with <15% RSD in soil and water, respectively. The limit of quantification (LOQ) was 0.0165 mg kg(-1) in soil and 0.132 μg L(-1) in water. Greater quantities of DDTs were extracted from soil using the MAE and SE techniques than with the SFE method. Higher amounts of DDTs were discovered in the southern (2.2-936 × 10(2) mg kg(-1)) or southwestern (86.3-2067 × 10(2) mg kg(-1)) direction from the factory than in the eastern direction (1.0-48.6 × 10(2) mg kg(-1)). An exception was the soil sample collected 50 ft (15.24 m) east (2904 × 10(2) mg kg(-1)) of the factory. The spread of DDTs in the water bodies (0.59-3.01 μg L(-1)) was approximately equal in all directions. We concluded that DDTs might have been dumped randomly around the warehouse after the closing of the factory.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Schwefelsäure, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Natriumsulfat, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Natriumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Natriumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumsulfat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Schwefelsäure, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Natriumsulfat, ≥99.0%, suitable for plant cell culture