Merck
  • Improved dissolution and absorption of ketoconazole in the presence of organic acids as pH-modifiers.
Improved dissolution and absorption of ketoconazole in the presence of organic acids as pH-modifiers.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-05-20)
Masashi Adachi, Yuta Hinatsu, Kosuke Kusamori, Hidemasa Katsumi, Toshiyasu Sakane, Manabu Nakatani, Koichi Wada, Akira Yamamoto
ZUSAMMENFASSUNG
Formulation development of poorly water-soluble compounds can be challenging because of incomplete dissolution that causes low and variable bioavailability. Enhancing compound solubility is important and many techniques have been investigated to that end, but they require specific materials and machinery. This study investigates the incorporation of a pH-modifier as a method to increase compound solubility and uses ketoconazole (KZ), which is weakly basic (pKa: 6.5), as a model compound. Organic acids are effective pH-modifiers and are generally used in pharmaceutical industries. We successfully obtained granules containing variable organic acids (KZ/acid granule) using a high-shear mixer. Dissolution tests of the KZ/acid granule resulted in highly enhanced solubility under non-sink conditions. Adding water-soluble acids, such as citric acid (CA) and tartaric acid, resulted in more than 8-fold higher dissolution at pH 6.0 compared to that of KZ only. The granules containing citric acid (KZ/CA granule) improved the dissolution of KZ after oral administration to rats under low gastric acid conditions, where the bioavailability of the KZ/CA granules at elevated gastric pH was comparable with that of KZ only at gastric acidic pH. The incorporation of organic acids would result in effective therapeutic outcomes independent of gastric pH in patients. In addition, higher bioavailability of KZ was observed after oral administration of KZ/CA granules under gastric acidic pH conditions than that of KZ alone. Thus, CA improved the dissolution and absorption rate of KZ after oral administration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Asparaginsäure, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Apfelsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Asparaginsäure, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Apfelsäure, 99%
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
L-Asparaginsäure, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
DL-Apfelsäure, ≥98% (capillary GC)
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
L-Asparaginsäure, BioUltra, ≥99.5% (T)
SAFC
L-Asparaginsäure
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Apfelsäure, meets USP/NF testing specifications
Sigma-Aldrich
DL-Apfelsäure, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)