Merck
The chemokine CCL20 induces proinflammatory and matrix degradative responses in cartilage.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2015-07-21)
Nada Alaaeddine, John Antoniou, Mayssam Moussa, George Hilal, Gaby Kreichaty, Ismat Ghanem, Wissam Abouchedid, Elie Saghbini, John A Di Battista
ZUSAMMENFASSUNG
Local inflammation plays a role in the pathophysiology of osteoarthritis (OA) and chemokines exert catabolic effects on articular cartilage either through paracrine and/or autocrine mechanisms. We sought to compare the expression levels of the chemokine (C-C motif) ligand 20 (CCL20) and its chemokine receptor 6 (CCR6) in donor and osteoarthritic (OA) cartilage and to investigate the role of CCL20 in the pathogenesis of OA and chondrocyte phenotype. Cartilage/chondrocytes from donor and OA knee joints was analyzed for CCL20 and CCR6 expression by RT-PCR and immunohistochemistry. Effects of CCL20 on cytokines and mediators of cartilage degradation were examined by RT-PCR for mRNA expression levels, enzyme-linked immunosorbent assays, and proteoglycan (GAG) assays. CCL20 and CCR6 proteins were abundantly expressed in OA cartilage sections compared to donor sections as judged by immunohistochemistry. RT-PCR of cartilage extracts confirmed the predominance of CCL20/CCR6 mRNA expression in OA cartilage. CCL20 mRNA expression was low in donor chondrocytes but increased after stimulation with proinflammatory cytokines. mRNA expression levels of IL-6, cyclooxygenase-2, and iNOS were elevated in donor chondrocyte cultures treated with rhCCL20. The release of MMP1/13, PGE2, proteoglycan GAG fragments, and IL-6 from cartilage explant cultures was markedly augmented in the presence of CCL-20. CCL-20 stimulated MMP-13, ADAMTS-5, and col type X mRNA but inhibited col type II mRNA expression in freshly explanted and cultured cartilage specimens. CCL20/CCR6 may play an important role in the pathogenesis of OA by inducing changes in phenotype and catabolic gene expression in chondrocytes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Cetylpyridiniumchlorid, meets USP testing specifications
Sigma-Aldrich
Tumornekrosefaktor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture