Merck
In Silico Estimation of Skin Concentration Following the Dermal Exposure to Chemicals.
Pharmaceutical research (2015-07-22)
Tomomi Hatanaka, Shun Yoshida, Wesam R Kadhum, Hiroaki Todo, Kenji Sugibayashi
ZUSAMMENFASSUNG
To develop an in silico method based on Fick's law of diffusion to estimate the skin concentration following dermal exposure to chemicals with a wide range of lipophilicity. Permeation experiments of various chemicals were performed through rat and porcine skin. Permeation parameters, namely, permeability coefficient and partition coefficient, were obtained by the fitting of data to two-layered and one-layered diffusion models for whole and stripped skin. The mean skin concentration of chemicals during steady-state permeation was calculated using the permeation parameters and compared with the observed values. All permeation profiles could be described by the diffusion models. The estimated skin concentrations of chemicals using permeation parameters were close to the observed levels and most data fell within the 95% confidence interval for complete prediction. The permeability coefficient and partition coefficient for stripped skin were almost constant, being independent of the permeant's lipophilicity. Skin concentration following dermal exposure to various chemicals can be accurately estimated based on Fick's law of diffusion. This method should become a useful tool to assess the efficacy of topically applied drugs and cosmetic ingredients, as well as the risk of chemicals likely to cause skin disorders and diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
4-Hydroxy-benzoesäure-ethylester, ReagentPlus®, 99%
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
4-Amino-benzoesäure-ethylester, ≥99% (HPLC)
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Butyl 4-hydroxybenzoat, ≥99.0% (GC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoesäure-propylester, ≥99%
Supelco
Methylparaben, PESTANAL®, analytical standard
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ethyl-4-aminobenzoat, 98%
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Benzoesäure, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Methyl-4-aminobenzoat, 98%
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid