Merck
  • Synthesis, DNA Binding, and Antiproliferative Activity of Novel Acridine-Thiosemicarbazone Derivatives.
Synthesis, DNA Binding, and Antiproliferative Activity of Novel Acridine-Thiosemicarbazone Derivatives.
International journal of molecular sciences (2015-06-13)
Sinara Mônica Vitalino de Almeida, Elizabeth Almeida Lafayette, Lúcia Patrícia Bezerra Gomes da Silva, Cézar Augusto da Cruz Amorim, Tiago Bento de Oliveira, Ana Lucia Tasca Gois Ruiz, João Ernesto de Carvalho, Ricardo Olímpio de Moura, Eduardo Isidoro Carneiro Beltrão, Maria do Carmo Alves de Lima, Luiz Bezerra de Carvalho Júnior
ZUSAMMENFASSUNG
In this work, the acridine nucleus was used as a lead-compound for structural modification by adding different substituted thiosemicarbazide moieties. Eight new (Z)-2-(acridin-9-ylmethylene)-N-phenylhydrazinecarbothioamide derivatives (3a-h) were synthesized, their antiproliferative activities were evaluated, and DNA binding properties were performed with calf thymus DNA (ctDNA) by electronic absorption and fluorescence spectroscopies. Both hyperchromic and hypochromic effects, as well as red or blue shifts were demonstrated by addition of ctDNA to the derivatives. The calculated binding constants ranged from 1.74 × 10(4) to 1.0 × 10(6) M(-1) and quenching constants from -0.2 × 10(4) to 2.18 × 10(4) M(-1) indicating high affinity to ctDNA base pairs. The most efficient compound in binding to ctDNA in vitro was (Z)-2-(acridin-9-ylmethylene)-N- (4-chlorophenyl) hydrazinecarbothioamide (3f), while the most active compound in antiproliferative assay was (Z)-2-(acridin-9-ylmethylene)-N-phenylhydrazinecarbothioamide (3a). There was no correlation between DNA-binding and in vitro antiproliferative activity, but the results suggest that DNA binding can be involved in the biological activity mechanism. This study may guide the choice of the size and shape of the intercalating part of the ligand and the strategic selection of substituents that increase DNA-binding or antiproliferative properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrazin Monohydrat, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazin Hydrat, reagent grade, N2H4 50-60 %
Sigma-Aldrich
2-Phenylethyl-isothiocyanat, FG
Sigma-Aldrich
Phenethylisothiocyanat, 99%