Merck
  • A Possible Freshness Marker for Royal Jelly: Formation of 5-Hydroxymethyl-2-furaldehyde as a Function of Storage Temperature and Time.
A Possible Freshness Marker for Royal Jelly: Formation of 5-Hydroxymethyl-2-furaldehyde as a Function of Storage Temperature and Time.
Journal of agricultural and food chemistry (2015-04-11)
Marco Ciulu, Ignazio Floris, Valeria M Nurchi, Angelo Panzanelli, Maria I Pilo, Nadia Spano, Gavino Sanna
ZUSAMMENFASSUNG
In this article we present a study of the variability of the concentration of 5-hydroxymethyl-2-furaldehyde (HMF) in natural royal jelly (RJ) as a function of its storage temperature (-18, 4, and 25 °C) and time (up to 9 months after harvesting). For this work HMF is evaluated using an RP-HPLC method we previously assessed. While all RJ samples stored at 4 and -18 °C always showed levels of HMF under the limit of detection (0.13 mg kg(-1)), samples kept at 25 °C showed an exponential increase in the concentration of HMF as a function of the storage time. This behavior and a number of desirable features of the analytical method used (ease of use in routine laboratories, availability of a complete validation protocol specifically developed for RJ, based on consolidated chemical knowledge) allow us to hypothesize the use of HMF as a possible, reliable freshness marker for RJ.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
Natriumtrichloracetat, 97%
Sigma-Aldrich
Trichloressigsäure, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.