Merck
  • Quantitative analysis of 17 amino acids in tobacco leaves using an amino acid analyzer and chemometric resolution.
Quantitative analysis of 17 amino acids in tobacco leaves using an amino acid analyzer and chemometric resolution.
Journal of separation science (2015-04-14)
Yihang Zeng, Wensheng Cai, Xueguang Shao
ZUSAMMENFASSUNG
A method was developed for quantifying 17 amino acids in tobacco leaves by using an A300 amino acid analyzer and chemometric resolution. In the method, amino acids were eluted by the buffer solution on an ion-exchange column. After reacting with ninhydrin, the derivatives of amino acids were detected by ultraviolet detection. Most amino acids are separated by the elution program. However, five peaks of the derivatives are still overlapping. A non-negative immune algorithm was employed to extract the profiles of the derivatives from the overlapping signals, and then peak areas were adopted for quantitative analysis of the amino acids. The method was validated by the determination of amino acids in tobacco leaves. The relative standard deviations (n = 5) are all less than 2.54% and the recoveries of the spiked samples are in a range of 94.62-108.21%. The feasibility of the method was proved by analyzing the 17 amino acids in 30 tobacco leaf samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
L-Isoleucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Buttersäure, ≥99%
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
L-Isoleucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
SAFC
Glycin
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
SAFC
L-Threonin
SAFC
L-Isoleucin
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
D-Serin, ≥98% (TLC)
Sigma-Aldrich
Buttersäure, ≥99%, FG
Sigma-Aldrich
DL-Asparaginsäure, ≥99% (TLC)
Sigma-Aldrich
L-Isoleucin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
DL-Phenylalanin, ReagentPlus®, 99%
Sigma-Aldrich
Buttersäure, natural, ≥99%, FCC, FG
Sigma-Aldrich
DL-Prolin, ReagentPlus®, 99%
Sigma-Aldrich
Glycin, 99%, FCC
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)