Merck
  • Are Neutrophil Extracellular Traps Playing a Role in the Parasite Control in Active American Tegumentary Leishmaniasis Lesions?
Are Neutrophil Extracellular Traps Playing a Role in the Parasite Control in Active American Tegumentary Leishmaniasis Lesions?
PloS one (2015-07-21)
Fernanda Nazaré Morgado, Michelle T C Nascimento, Elvira M Saraiva, Carla de Oliveira-Ribeiro, Maria de Fátima Madeira, Marcela da Costa-Santos, Erica C F Vasconcellos, Maria Ines F Pimentel, Marcelo Rosandiski Lyra, Armando de Oliveira Schubach, Fátima Conceição-Silva
ZUSAMMENFASSUNG
Neutrophil extracellular traps (NETs) have been described as a network of extracellular fibers composed by DNA, histones and various proteins/enzymes. Studies have demonstrated that NETs could be responsible for the trapping and elimination of a variety of infectious agents. In order to verify the presence of NETs in American tegumentary leishmaniasis (ATL) and their relationship with the presence of amastigotes we evaluated active cutaneous lesions of 35 patients before treatment by the detection of parasites, neutrophils (neutrophil elastase) and histones through immunohistochemistry and confocal immunofluorescence. Intact neutrophils could be detected in all ATL lesions. NETs were present in 27 patients (median 1.1; range from 0.1 to 23.5/mm2) with lesion duration ranging from one to seven months. NETs were in close proximity with neutrophils (r = 0.586; p = 0.0001) and amastigotes (r = 0.710; p = 0.0001). Two patterns of NET formation were detected: small homogeneously distributed networks observed in all lesions; and large structures that could be visualized at a lower magnification in lesions presenting at least 20% of neutrophils. Lesions presenting the larger NET formation showed high parasite detection. A correlation between NET size and the number of intact amastigotes was observed (p=0.02). As we detected an association between NET and amastigotes, our results suggest that neutrophil migration and NET formation could be stimulated and maintained by stimuli derived from the parasite burden/parasite antigen in the extracellular environment. The observation of areas containing only antigens not intermingled with NETs (elastase and histone) suggests that the involvement of these structures in the control of parasite burden is a dynamic process in which the formation of NETs is exhausted with the destruction of the parasites. Since NETs were also associated with granulomas, this trapping would favor the activity of macrophages in order to control the parasite burden.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC