Merck
  • New strategy for expression of recombinant hydroxylated human collagen α1(III) chains in Pichia pastoris GS115.
New strategy for expression of recombinant hydroxylated human collagen α1(III) chains in Pichia pastoris GS115.
Biotechnology and applied biochemistry (2014-06-24)
Jing He, Xiaoxuan Ma, Fenglong Zhang, Linbo Li, Jianjun Deng, Wenjiao Xue, Chenhui Zhu, Daidi Fan
ZUSAMMENFASSUNG
Type III collagen is one of the most abundant proteins in the human body, which forms collagen fibrils and provides the stiff, resilient characteristics of many tissues. In this paper, a new method for secretory expression of recombinant hydroxylated human collagen α1(III) chain in Pichia pastoris GS115 was applied. The gene encoding for full-length human collagen α1(III) chain (COL3A1) without N-terminal propeptide and C-terminal propeptide was cloned in the pPIC9K expression vector. The prolyl 4-hydroxylase (P4H, EC 1.14.11.2) α-subunit (P4Hα) and β-subunit (P4Hβ) genes were cloned in the same expression vector, pPICZB. Fluorogenic quantitative PCR indicates that COL3A1 and P4H genes have been expressed in mRNA level. SDS-PAGE shows that secretory expression of recombinant human collagen α1(III) chain was successfully achieved in P. pastoris GS115. In addition, the result of amino acids composition analysis shows that the recombinant human collagen α1(III) chain contains hydroxyproline by coexpression with the P4H. Furthermore, liquid chromatography coupled with tandem mass spectrometry analysis demonstrates that proline residues of the recombinant human collagen α1(III) chain were hydroxylated in the X or Y positions of Gly-X-Y triplets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, γ-irradiated, powder, BioXtra, suitable for cell culture