Merck
  • Chitoporin from the Marine Bacterium Vibrio harveyi: PROBING THE ESSENTIAL ROLES OF TRP136 AT THE SURFACE OF THE CONSTRICTION ZONE.
Chitoporin from the Marine Bacterium Vibrio harveyi: PROBING THE ESSENTIAL ROLES OF TRP136 AT THE SURFACE OF THE CONSTRICTION ZONE.
The Journal of biological chemistry (2015-06-18)
Watcharin Chumjan, Mathias Winterhalter, Albert Schulte, Roland Benz, Wipa Suginta
ZUSAMMENFASSUNG
VhChiP is a sugar-specific porin present in the outer membrane of the marine bacterium Vibrio harveyi. VhChiP is responsible for the uptake of chitin oligosaccharides, with particular selectivity for chitohexaose. In this study, we employed electrophysiological and biochemical approaches to demonstrate that Trp(136), located at the mouth of the VhChiP pore, plays an essential role in controlling the channel's ion conductivity, chitin affinity, and permeability. Kinetic analysis of sugar translocation obtained from single channel recordings indicated that the Trp(136) mutations W136A, W136D, W136R, and W136F considerably reduce the binding affinity of the protein channel for its best substrate, chitohexaose. Liposome swelling assays confirmed that the Trp(136) mutations decreased the rate of bulk chitohexaose permeation through the VhChiP channel. Notably, all of the mutants show increases in the off-rate for chitohexaose of up to 20-fold compared with that of the native channel. Furthermore, the cation/anion permeability ratio Pc/Pa is decreased in the W136R mutant and increased in the W136D mutant. This demonstrates that the negatively charged surface at the interior of the protein lumen preferentially attracts cationic species, leading to the cation selectivity of this trimeric channel.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid -Lösung, ~30% in H2O
Sigma-Aldrich
Hexadecan, ReagentPlus®, 99%
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hexadecan, anhydrous, ≥99%
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer