Merck
  • Adolescent testosterone influences BDNF and TrkB mRNA and neurotrophin-interneuron marker relationships in mammalian frontal cortex.
Adolescent testosterone influences BDNF and TrkB mRNA and neurotrophin-interneuron marker relationships in mammalian frontal cortex.
Schizophrenia research (2015-06-20)
Tertia D Purves-Tyson, Katherine Allen, Samantha Fung, Debora Rothmond, Pam L Noble, David J Handelsman, Cynthia Shannon Weickert
ZUSAMMENFASSUNG
Late adolescence in males is a period of increased susceptibility for the onset of schizophrenia, coinciding with increased circulating testosterone. The cognitive deficits prevalent in schizophrenia may be related to unhealthy cortical interneurons, which are trophically dependent on brain derived neurotrophic factor. We investigated, under conditions of depleted (monkey and rat) and replaced (rat) testosterone over adolescence, changes in gene expression of cortical BDNF and TrkB transcripts and interneuron markers and the relationships between these mRNAs and circulating testosterone. Testosterone removal by gonadectomy reduced gene expression of some BDNF transcripts in monkey and rat frontal cortices and the BDNF mRNA reduction was prevented by testosterone replacement. In rat, testosterone replacement increased the potential for classical TrkB signalling by increasing the full length to truncated TrkB mRNA ratio, whereas in the monkey cortex, circulating testosterone was negatively correlated with the TrkB full length/truncated mRNA ratio. We did not identify changes in interneuron gene expression in monkey frontal cortex in response to gonadectomy, and in rat, we showed that only somatostatin mRNA was decreased by gonadectomy but not restored by testosterone replacement. We identified complex and possibly species-specific, relationships between BDNF/TrkB gene expression and interneuron marker gene expression that appear to be dependent on the presence of testosterone at adolescence in rat and monkey frontal cortices. Taken together, our findings suggest there are dynamic relationships between BDNF/TrkB and interneuron markers that are dependent on the presence of testosterone but that this may not be a straightforward increase in testosterone leading to changes in BDNF/TrkB that contributes to interneuron health.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri