Merck
  • Rational combination of MEK inhibitor and the STAT3 pathway modulator for the therapy in K-Ras mutated pancreatic and colon cancer cells.
Rational combination of MEK inhibitor and the STAT3 pathway modulator for the therapy in K-Ras mutated pancreatic and colon cancer cells.
Oncotarget (2015-05-12)
Chengguang Zhao, Hui Xiao, Xiaojuan Wu, Chenglong Li, Guang Liang, Shulin Yang, Jiayuh Lin
ZUSAMMENFASSUNG
K-Ras mutations are frequently detected in pancreatic and colon cancers, which are associated with the resistance to MEK inhibitors targeting the Ras pathway. Identifying the underlying mechanisms for the acquired resistance is essential for the future clinical development of MEK inhibitors. Here, we identified that Signal Transducer and Activator of Transcription 3 (STAT3) was significantly activated following the MEK inhibition using AZD6244, PD98059 and Trametinib in K-Ras mutant pancreatic and colon cancer cells. The STAT3 activation may be important for the MEK inhibitor resistance in these K-Ras mutant cancer cells. We have shown that dual inhibition of STAT3 and MEK using the STAT3 inhibitor LY5 and MEK inhibitor Trametinib exerts significant anti-tumor cell efficacy in K-Ras mutant pancreatic and colon cancer cells in vitro. In addition, Trametinib showed increased suppression on tumor growth in vivo in STAT3 knockdown pancreatic cancer cells compared with tumor growth of control cells without STAT3 knockdown. Taken together, our results suggest the induced STAT3 activation as a possible mechanism for the resistance to MEK inhibitor and demonstrate the potentials of a combination therapy using MEK and STAT3 inhibitors in pancreatic and colon cancers harboring K-Ras mutant proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Glutamin
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
SAFC
L-Threonin
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
2-Butanon, FCC, FG
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)