Merck
  • Cylindromatosis (CYLD) inhibits Streptococcus pneumonia-induced plasminogen activator inhibitor-1 expression via interacting with TRAF-6.
Cylindromatosis (CYLD) inhibits Streptococcus pneumonia-induced plasminogen activator inhibitor-1 expression via interacting with TRAF-6.
Biochemical and biophysical research communications (2015-06-10)
Jing Wu, Jing Zhao, Jie Yu, Wenhong Zhang, Yuxian Huang
ZUSAMMENFASSUNG
Streptococcus pneumoniae (S. p) remains one of the foremost causes of community-acquired pneumonia. Recent studies have shown that S. p lung infection is associated with plasminogen activator inhibitor-1 (PAI-1) expression, which inhibits acute lung injury. Such effects by S. p were negatively regulated by cylindromatosis (CYLD). The current study explored the underlying mechanisms. We showed that S. p-induced PAI-1 expression requires tumor necrosis factor receptor-associated factor 6 (TRAF-6) signaling. Si-RNA-mediated knockdown of TRAF-6 remarkably inhibited S. p-induced PAI-1 expression. Reversely, over-expression of wild type (wt-) TRAF-6 further potentiated PAI-1 expression in S. p-treated cells. We provided evidences to support that CYLD-mediated anti-PAI-1 activity might be through direct regulation of TRAF-6. Our results from co-immunoprecipitation (co-IP) and confocal microscopy assays confirmed a direct association between the CYLD and TRAF-6 in A549 cells. Over-expression of wt-CYLD remarkably inhibited TRAF-6 ubiquitination and subsequent PAI-1 expression. Introducing a mutated CYLD, on the other hand, enhanced TRAF-6 ubiquitination and PAI-1 expression. Together, these results indicate that TRAF-6 mediates S. p-induced PAI-1 expression, and CYLD inhibits PAI-1 expression probably through deubiquitinating TRAF-6. The current study provided molecular insights of CYLD-mediated activities in S. p-induced PAI-1 expression and possible acute lung injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid