Merck
  • Self nanoemulsifying drug delivery system of stabilized ellagic acid-phospholipid complex with improved dissolution and permeability.
Self nanoemulsifying drug delivery system of stabilized ellagic acid-phospholipid complex with improved dissolution and permeability.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society (2015-06-25)
Amelia M Avachat, Vijay G Patel
ZUSAMMENFASSUNG
Ellagic acid (EA), a plant polyphenol known for its wide-range of health benefits has limited use due to its low oral bioavailability. In this study, a new self-nanoemulsifying drug delivery system (SNEDDS), based on the phospholipid complex technique, was developed to improve the oral bioavailability of ellagic acid. Ellagic acid-phospholipid complex was prepared by an anti-solvent method and characterized. Enhanced lipophilicity after the formation of ellagic acid-phospholipid complex was verified through solubility studies. Preliminary screening was carried out to select oil, surfactant and co-surfactant. Ternary phase diagrams were constructed to identify the area of nanoemulsification. Formulations were optimized on the basis of globule size, cloud point and robustness to dilution. The optimized SNEDDS of ellagic acid-phospholipid complex showed mean globule size of 106 ± 0.198 nm and cloud point at 83-85 °C. The in vitro drug release from SNEDDS was found to be higher compared to EA suspension and complex, while ex vivo studies showed increased permeation from SNEDDS compared to EA suspension. Moreover, SNEDDS overcome the food effect which was shown by EA suspension. Thus, SNEDDS were found to be influential in improving the release performance of EA, indicating their potential to improve the oral bioavailability of EA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Kaliumbromid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Kaliumbromid, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
Kaliumbromid, BioXtra, ≥99.0%