Merck
From Soybean residue to advanced supercapacitors.
Scientific reports (2015-11-17)
G A Ferrero, A B Fuertes, M Sevilla
ZUSAMMENFASSUNG
Supercapacitor technology is an extremely timely area of research with fierce international competition to develop cost-effective, environmentally friendlier EC electrode materials that have real world application. Herein, nitrogen-doped carbons with large specific surface area, optimized micropore structure and surface chemistry have been prepared by means of an environmentally sound hydrothermal carbonization process using defatted soybean (i.e., Soybean meal), a widely available and cost-effective protein-rich biomass, as precursor followed by a chemical activation step. When tested as supercapacitor electrodes in aqueous electrolytes (i.e. H2SO4 and Li2SO4), they demonstrate excellent capacitive performance and robustness, with high values of specific capacitance in both gravimetric (250-260 and 176 F g(-1) in H2SO4 and Li2SO4 respectively) and volumetric (150-210 and 102 F cm(-3) in H2SO4 and Li2SO4 respectively) units, and remarkable rate capability (>60% capacitance retention at 20 A g(-1) in both media). Interestingly, when Li2SO4 is used, the voltage window is extended up to 1.7 V (in contrast to 1.1 V in H2SO4). Thus, the amount of energy stored is increased by 50% compared to H2SO4 electrolyte, enabling this environmentally sound Li2SO4-based supercapacitor to deliver ~12 Wh kg(-1) at a high power density of ~2 kW kg(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Kaliumhydroxid, technical, ≥85%, powder
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, >40 μm particle size
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Poly(tetrafluorethylen), beads
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Poly(tetrafluorethylen), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluorethylen), powder, ≥350 μm particle size
Sigma-Aldrich
Kaliumhydroxid, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets