Merck
  • First proteome study of sporadic flowering in bamboo species (Bambusa vulgaris and Dendrocalamus manipureanus) reveal the boom is associated with stress and mobile genetic elements.
First proteome study of sporadic flowering in bamboo species (Bambusa vulgaris and Dendrocalamus manipureanus) reveal the boom is associated with stress and mobile genetic elements.
Gene (2015-08-12)
Bengyella Louis, Sayanika Devi Waikhom, Sailendra Goyari, Robinson C Jose, Pranab Roy, Narayan Chandra Talukdar
ZUSAMMENFASSUNG
Bamboo species are the fastest-growing plants having a long vegetative cycle. Abrupt switching from the vegetative phase to the reproductive phase via sporadic flowering boom, occasionally leads to death of bamboo clumps, and threatens the existence of many bamboo species. To apprehend the molecular mechanism driving sporadic flowering, proteome changes in the initial and advanced floral buds of two edible bamboo species (Bambusa vulgaris and Dendrocalamus manipureanus) was dissected by two-dimensional gel electrophoresis (2-DE). A total of 39 differentially expressed peptide spots were identified by matrix-assisted laser desorption ionization time-of-flight/time-of-flight mass spectrometry (MALDI-TOF-TOF/MS). In both B. vulgaris and D. manipureanus, identified proteins were categorized as transposon-related, defence and stress-related, cell cycle related, metabolism related, signal transduction related, and some lacked known putative domains. Proteins such as SEPALLATA3, ubiquitin, histone 3, thaumatin-like protein, putative tethering factor, SF-assemblin, polyubiquitin, mitochondrial carrier-like protein and RPT2-like protein were significantly expressed. Differences in D. manipureanus and B. vulgaris suggested that bamboo species have diverse 'drivers' or 'passengers' genes that govern natural sporadic flowering boom. This first floral proteomics analysis of bamboos revealed that sporadic boom is a highly energetic process, associated with stress elements, mobile genetic elements and signal transduction cross-talk elements.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Bromphenolblau, titration: suitable
SAFC
Glycin
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)