Merck
  • Production of biodiesel from vegetable oil and microalgae by fatty acid extraction and enzymatic esterification.
Production of biodiesel from vegetable oil and microalgae by fatty acid extraction and enzymatic esterification.
Journal of bioscience and bioengineering (2015-01-13)
Beatriz Castillo López, Luis Esteban Cerdán, Alfonso Robles Medina, Elvira Navarro López, Lorena Martín Valverde, Estrella Hita Peña, Pedro A González Moreno, Emilio Molina Grima
ZUSAMMENFASSUNG
The aim of this work was to obtain biodiesel (methyl esters) from the saponifiable lipids (SLs) fraction of the microalga Nannochloropsis gaditana, whose biomass dry weight contains 12.1 wt% of these lipids. SLs were extracted from the microalga as free fatty acids (FFAs) for subsequent transformation to methyl esters (biodiesel) by enzymatic esterification. Extraction as FFAs rather than as SLs allows them to be obtained with higher purity. Microalgal FFAs were obtained by direct saponification of lipids in the biomass and subsequent extraction-purification with hexane. Esterification of FFAs with methanol was catalysed by lipase Novozym 435 from Candida antarctica. Stability studies of this lipase in the operational conditions showed that the esterification degree (ED) attained with the same batch of lipase remained constant over six reaction cycles (36 h total reaction time). The optimal conditions attained for 4 g of FFAs were 25°C, 200 rpm, methanol/FFA molar ratio of 1.5:1, Novozym 435/FFA ratio of 0.025:1 w/w and 4 h reaction time. In these conditions the ED attained was 92.6%, producing a biodiesel with 83 wt% purity from microalgal FFAs. Several experimental scales were tested (from 4 to 40 g FFAs), and in all cases similar EDs were obtained.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Nonadecansäure, ≥98% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O