Merck
  • Se-Methyl-L-selenocysteine Induces Apoptosis via Endoplasmic Reticulum Stress and the Death Receptor Pathway in Human Colon Adenocarcinoma COLO 205 Cells.
Se-Methyl-L-selenocysteine Induces Apoptosis via Endoplasmic Reticulum Stress and the Death Receptor Pathway in Human Colon Adenocarcinoma COLO 205 Cells.
Journal of agricultural and food chemistry (2015-05-07)
Yen-Chen Tung, Mei-Ling Tsai, Fang-Ling Kuo, Ching-Shu Lai, Vladimir Badmaev, Chi-Tang Ho, Min-Hsiung Pan
ZUSAMMENFASSUNG
Selenomethionine (SeMet) and Se-methyl-L-selenocysteine (MSeC) are natural organoselenium compounds found in garlic, onion, and broccoli. In addition, these compounds have lower toxicity and better anticancer activities than inorganic Se. This study investigated the effects of MSeC treatment on the growth of COLO 205 human colorectal adenocarcinoma cells and evaluated the mechanisms related to the MSeC-induced effects. When COLO 205 cells were treated with 200 μM MSeC for 24 h, MSeC caused 80% apoptosis in cells. MSeC increased the expression of Fas and FasL, followed by the cleavage of caspase-3, caspase-8, DNA fragmentation factor (DFF45), and poly(ADP-ribose) polymerase (PARP). MSeC also increased the levels of Bax protein and decreased the levels of Bid and Bcl-2 proteins. However, MSeC did not cause apoptosis through reactive oxygen species (ROS) stress but instead through endoplasmic reticulum (ER) stress. The cleavage of caspase-12 and caspase-9 was shown to increase the growth arrest and protein levels of DNA-damage inducible genes (GADD) 153 and 45. MSeC also downregulated the ERK1/2 and PI3K/AKT protein levels and upregulated the p38 and JNK protein levels in COLO 205 cells. These results showed that the mechanism by which MSeC induced apoptosis in COLO 205 cells involved caspase activation, the extrinsic apoptotic pathway, and the regulation of ER-stress-induced apoptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
SAFC
Glycin
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Glycin, 99%, FCC