Merck
  • Extraction optimization, characterization and antioxidant activity in vitro of polysaccharides from mulberry (Morus alba L.) leaves.
Extraction optimization, characterization and antioxidant activity in vitro of polysaccharides from mulberry (Morus alba L.) leaves.
Carbohydrate polymers (2015-05-26)
Qingxia Yuan, Yufeng Xie, Wei Wang, Yuhua Yan, Hong Ye, Saqib Jabbar, Xiaoxiong Zeng
ZUSAMMENFASSUNG
Extraction optimization, characterization and antioxidant activity in vitro of polysaccharides from mulberry leaves (MLP) were investigated in the present study. The optimal extraction conditions with an extraction yield of 10.0 ± 0.5% for MLP were determined as follows: extraction temperature 92 °C, extraction time 3.5h and ratio (v/w, mL/g) of extraction solvent (water) to raw material 34. Two purified fractions, MLP-3a and MLP-3b with molecular weights of 80.99 and 3.64 kDa, respectively, were obtained from crude MLP by chromatography of DEAE-Cellulose 52 and Sephadex G-100. Fourier transform-infrared spectroscopy revealed that crude MLP, MLP-3a and MLP-3b were acidic polysaccharides. Furthermore, crude MLP and MLP-3a had more complicated monosaccharide compositions, while MLP-3b had a relatively higher content of uronic acid. Crude MLP, MLP-3a and MLP-3b exhibited potent Fe(2+) chelating power and scavenging activities on 1,1-diphenyl-2-picrylhydrazyl, hydroxyl, superoxide and 2,2'-azinobis-(3-ethyl-benzothiazolin-6-sulfonic acid) radicals. The results suggested that MLP could be explored as natural antioxidant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-Glucuronsäure, ≥98% (GC)
Supelco
2,4,6-Tri-(2-pyridyl)-s-triazin, ≥98%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Phenazin-Methosulfat
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
Phenazin, 98%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazin, 97%
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP