Merck
  • Sono-intercalation of CdS nanoparticles into the layers of titanate facilitates the sunlight degradation of Congo red.
Sono-intercalation of CdS nanoparticles into the layers of titanate facilitates the sunlight degradation of Congo red.
Journal of colloid and interface science (2015-10-11)
S Sehati, M H Entezari
ZUSAMMENFASSUNG
In this paper, the degradation of Congo red (CR) as a dye was investigated by a new synthetic photocatalyst. The synthesis was done through the intercalation of CdS in the layers of titanate (K2Ti4O9) by the assistance of ultrasound. The photocatalyst was prepared via ion-exchange reaction and sulfuration processes in the presence of ultrasonic irradiation. The samples were characterized by the field emission scanning electron microscopy (FESEM), transmission electron microscopy (TEM), X-ray diffraction (XRD), ultraviolet-visible spectra (UV-Vis), and Fourier transform infrared spectroscopy (FTIR). The deposition of CdS nanoparticles on the surface and between the spaces of titanate layers led to the shift of absorption edge of titanate to the visible light region. The photocatalytic degradation mechanism of CR has been evaluated through the addition of some scavengers to the solution. In addition, the stability and reusability of the catalyst were examined in this work too.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Kaliumiodid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Kaliumiodid, ReagentPlus®, 99%
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Ammoniak -Lösung, 7 N in methanol
Sigma-Aldrich
Kaliumpersulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Kaliumiodid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Butylamin, 99.5%
Sigma-Aldrich
Ammoniak -Lösung, 2.0 M in ethanol
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%