Merck
  • Effect of sodium/iodide symporter (NIS)-mediated radioiodine therapy on estrogen receptor-negative breast cancer.
Effect of sodium/iodide symporter (NIS)-mediated radioiodine therapy on estrogen receptor-negative breast cancer.
Oncology reports (2015-05-09)
Chen Yao, Yi Pan, Yongxin Li, Xiangdong Xu, Ying Lin, Wenjian Wang, Shenming Wang
ZUSAMMENFASSUNG
Since the sodium/iodide symporter (NIS) stimulates the iodine uptake in normal lactating breast, our study aimed to study the effect of NIS-mediated radioiodide therapy on ER-negative breast cancers. A recombinant lentivirus plasmid encoding the human NIS (hNIS) gene and firefly luciferase (Fluc) was constructed. MDA-MB-231 cells were transfected with the recombinant lentivirus, and the hNIS gene expression was identified by western blot analysis and real-time PCR. Tissue-specific expression of the NIS gene was confirmed by immunohistochemical (IHC) staining. Functional NIS activity in the MDA-hNIS cells was confirmed by the uptake of 131I and cytotoxicity assays. The relative expression level of hNIS mRNA exhibited a 10-fold higher expression in the MDA-hNIS cells compared with the level in the control cells without the endogenous NIS gene. Abundant expression of hNIS protein was noted in the cell membrane compared to the cytoplasm which confirmed the efficient expression of the functional hNIS gene. Iodine uptake into the MDA-hNIS cells was rapid, reaching a maximum after 15 min, followed by a decline. Exposure of the MDA-hNIS cells with 131I resulted in a time-dependent reduction in colony formation compared with the survival of the control (MDA) cells. Our results confirmed that NIS overexpression enhances the sensitivity of ER-negative breast cancer cells to radioiodide therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Iodsuccinimid, 95%
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O