Merck
  • Genetic Deletion of β-Arrestin-2 and the Mitigation of Established Airway Hyperresponsiveness in a Murine Asthma Model.
Genetic Deletion of β-Arrestin-2 and the Mitigation of Established Airway Hyperresponsiveness in a Murine Asthma Model.
American journal of respiratory cell and molecular biology (2015-01-09)
Minyong Chen, Akhil Hegde, Yeon Ho Choi, Barbara S Theriot, Richard T Premont, Wei Chen, Julia K L Walker
ZUSAMMENFASSUNG
β-Arrestin-2 (βarr2) is a ubiquitously expressed cytosolic protein that terminates G protein-coupled receptor signaling and transduces G protein-independent signaling. We previously showed that mice lacking βarr2 do not develop an asthma phenotype when sensitized to, and challenged with, allergens. The current study evaluates if an established asthma phenotype can be mitigated by deletion of βarr2 using an inducible Cre recombinase. We sensitized and challenged mice to ovalbumin (OVA) and demonstrated that on Day (d) 24 the allergic asthma phenotype was apparent in uninduced βarr2 and wild-type (WT) mice. In a second group of OVA-treated mice, tamoxifen was injected on d24 to d28 to activate Cre recombinase, and OVA aerosol challenge was continued through d44. The asthma phenotype was assessed using lung mechanics measurements, bronchoalveolar lavage cell analysis, and histological assessment of mucin and airway inflammation. Compared with their respective saline-treated controls, OVA-treated WT mice and mice expressing the inducible Cre recombinase displayed a significant asthma phenotype at d45. Whereas tamoxifen treatment had no significant effect on the asthma phenotype in WT mice, it inhibited βarr2 expression and caused a significant reduction in airway hyper-responsiveness (AHR) in Cre-inducible mice. These findings suggest that βarr2 is actively required for perpetuation of the AHR component of the allergic asthma phenotype. Our finding that βarr2 participates in the perpetuation of AHR in an asthma model means that targeting βarr2 may provide immediate and potentially long-term relief from daily asthma symptoms due to AHR irrespective of inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%