Merck
  • Striatal-enriched protein tyrosine phosphatase controls responses to aversive stimuli: implication for ethanol drinking.
Striatal-enriched protein tyrosine phosphatase controls responses to aversive stimuli: implication for ethanol drinking.
PloS one (2015-05-21)
Rémi Legastelois, Emmanuel Darcq, Scott A Wegner, Paul J Lombroso, Dorit Ron
ZUSAMMENFASSUNG
The STriatal-Enriched protein tyrosine Phosphatase (STEP) is a brain-specific phosphatase whose dysregulation in expression and/or activity is associated with several neuropsychiatric disorders. We recently showed that long-term excessive consumption of ethanol induces a sustained inhibition of STEP activity in the dorsomedial striatum (DMS) of mice. We further showed that down-regulation of STEP expression in the DMS, and not in the adjacent dorsolateral striatum, increases ethanol intake, suggesting that the inactivation of STEP in the DMS contributes to the development of ethanol drinking behaviors. Here, we compared the consequence of global deletion of the STEP gene on voluntary ethanol intake to the consumption of an appetitive rewarding substance (saccharin) or an aversive solution (quinine or denatonium). Whereas saccharin intake was similar in STEP knockout (KO) and wild type (WT) littermate mice, the consumption of ethanol as well as quinine and denatonium was increased in STEP KO mice. These results suggested that the aversive taste of these substances was masked upon deletion of the STEP gene. We therefore hypothesized that STEP contributes to the physiological avoidance towards aversive stimuli. To further test this hypothesis, we measured the responses of STEP KO and WT mice to lithium-induced conditioned place aversion (CPA) and found that whereas WT mice developed lithium place aversion, STEP KO mice did not. In contrast, conditioned place preference (CPP) to ethanol was similar in both genotypes. Together, our results indicate that STEP contributes, at least in part, to the protection against the ingestion of aversive agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Denatoniumbenzoat, ≥98%
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Chinin, 90%
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Chinin, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithiumchlorid, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium-7Li-chlorid, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
N6-Cyclopentyladenosine, solid