Merck
Syntheses and crystal structures of vanadium and iron chloride complexes with diglyme.
Acta chimica Slovenica (2015-06-19)
Saša Petriček, Alojz Demšar
ZUSAMMENFASSUNG
A mononuclear molecular complex fac-[VCl3(diglyme)] (1) resulted from the reaction of VCl3 and diglyme (diglyme = di(2-methoxyethyl)ether) in dichloromethane. The violet complex 1 is a sensitive substance which slowly oxidized to a new, blue mononuclear molecular complex, fac-[VOCl2(diglyme)] (2) in the presence of air. The synthesis of iron(II), iron(III) complex [FeCl(diglyme)(THF)]2[FeCl4)]2 (3) was achieved by the reaction of yellow-green, partly oxidized FeCl2.4H2O, diglyme and chlorotrimethyl-silane in tetrahydofuran. The compound consists of the dinuclear cations with octahedral environment of iron(II) and the tetrahedral anions of iron(III). A pure iron(II) chloride-diglyme complex [FeCl2(diglyme)]2 (4) was gained by the reaction of freshly prepared iron(II) chloride hydrate, diglyme and chlorotrimethylsilane in dichloromethane. Diglyme is coordinated in the meridional mode to octahedral iron(II) in dinuclear cations of 3 and in dinuclear molecules 4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Vanadium(III)-chlorid, 97%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Chlorotrimethylsilan, purified by redistillation, ≥99%
Sigma-Aldrich
Diethylenglycoldimethylether, anhydrous, 99.5%
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chlorotrimethylsilan, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Chlortrimethylsilan -Lösung, 1.0 M in THF