Merck
  • Structure of an Amino Acid-Decorated Exopolysaccharide Secreted by a Vibrio alginolyticus Strain.
Structure of an Amino Acid-Decorated Exopolysaccharide Secreted by a Vibrio alginolyticus Strain.
Marine drugs (2015-11-04)
Sophie Drouillard, Isabelle Jeacomine, Laurine Buon, Claire Boisset, Anthony Courtois, Bertrand Thollas, Pierre-Yves Morvan, Romuald Vallée, William Helbert
ZUSAMMENFASSUNG
Vibrio alginolyticus (CNCM I-4994) secretes an exopolysaccharide that can be used as an ingredient in cosmetic applications. The structure was resolved using chromatography and one- and two-dimensional NMR spectroscopy experiments. The results show that the carbohydrate backbone is made of two residues: d-galacturonic acid and N-acetyl-d-glucosamine (GlcNac), which together constitute a tetrasaccharide repetition unit: [→3)-α-d-GalA-(1→4)-α-d-GalA-(1→3)-α-d-GalA-(1→3)-β-GlcNAc(1→]. Two amino acids, alanine and serine, are linked to GalA residues via amido linkages. The position and the distribution of the amino acids were characterized by two-dimensional NMR spectroscopy. To our knowledge, this is the first description of a structure for a marine exopolysaccharide decorated with an amino acid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Supelco
N,O-Bis-(trimethylsilyl)-trifluoracetamid mit Trimethylchlorsilan, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥99%
Sigma-Aldrich
Chlorotrimethylsilan, purified by redistillation, ≥99%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochinon, ReagentPlus®, 99%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Sigma-Aldrich
Chlorotrimethylsilan, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Chlortrimethylsilan -Lösung, 1.0 M in THF
Sigma-Aldrich
Hydrochinon, meets USP testing specifications