Merck
  • Expression of 15-Hydroxyprostaglandin Dehydrogenase in Human Chorion Is Associated with Peroxisome Proliferator-Activated Receptor Isoform Expression in Term Labor.
Expression of 15-Hydroxyprostaglandin Dehydrogenase in Human Chorion Is Associated with Peroxisome Proliferator-Activated Receptor Isoform Expression in Term Labor.
The American journal of pathology (2015-06-22)
Ping He, Yuan Li, Xiaoying Ding, Qianqian Sun, Ying Huang, Hang Gu, Xin Ni
ZUSAMMENFASSUNG
Chorionic NAD-dependent 15-hydroxy prostaglandin dehydrogenase (PGDH) plays a pivotal role in controlling the amount of prostaglandins in the uterus. Peroxisome proliferator-activated receptors (PPARs) are implicated to be involved in parturition. In this study, we investigated whether PPARs are involved in control of PGDH expression in chorion. The chorionic tissues were collected from the following groups of the women with singleton pregnancy: term no labor (TNL), term labor (TL) and preterm labor (PTL). Chorionic trophoblasts were isolated and cultured in vitro. Immunocytochemistry analysis showed that PPARα, PPARβ, and PPARγ were localized to trophoblasts in chorion. The protein levels of PGDH, PPARβ, and PPARγ were localized to trophoblasts in chorion. The protein levels of PPARα, PPARβ, and PPARγ were reduced in TL tissues compared to that of TNL group. PPARα, PPARβ, and PPARγ expression correlated to PGDH in TNL tissues, whereas only PPARγ expression correlated to PGDH in TL chorion tissues. PGDH expression was decreased in PTL tissues compared with TL group, whereas the expression of PPARs was not significantly different between TL and PTL groups. The agonists of three PPARs dose-dependently stimulated PGDH activity, mRNA, and protein expression in cultured chorionic cells. PPARs did not affect the stability of PGDH mRNA but stimulated the transcriptional activity of HPGD gene. Our results suggest that PPARs play pivotal roles in maintenance of PGDH expression in chorion during human pregnancy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Rosiglitazon
Sigma-Aldrich
Ethanolamin, ≥99%
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Ethanolamin, ≥98%
Sigma-Aldrich
Ethanolamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
Ethanolamin, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamin, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
GW9662, >98% (HPLC)
Sigma-Aldrich
GW7647, ≥98% (HPLC)