Merck
  • Evaluation of iTRAQ and SWATH-MS for the Quantification of Proteins Associated with Insulin Resistance in Human Duodenal Biopsy Samples.
Evaluation of iTRAQ and SWATH-MS for the Quantification of Proteins Associated with Insulin Resistance in Human Duodenal Biopsy Samples.
PloS one (2015-05-08)
Sylvie Bourassa, Frédéric Fournier, Benjamin Nehmé, Isabelle Kelly, André Tremblay, Valéry Lemelin, Benoit Lamarche, Patrick Couture, Arnaud Droit
ZUSAMMENFASSUNG
Insulin resistance (IR) is associated with increased production of triglyceride-rich lipoproteins of intestinal origin. In order to assess whether insulin resistance affects the proteins involved in lipid metabolism, we used two mass spectrometry based quantitative proteomics techniques to compare the intestinal proteome of 14 IR patients to that of 15 insulin sensitive (IS) control patients matched for age and waist circumference. A total of 3886 proteins were identified by the iTRAQ (Isobaric Tags for Relative and Absolute Quantitation) mass spectrometry approach and 2290 by the SWATH-MS strategy (Serial Window Acquisition of Theoretical Spectra). Using these two methods, 208 common proteins were identified with a confidence corresponding to FDR < 1%, and quantified with p-value < 0.05. The quantification of those 208 proteins has a Pearson correlation coefficient (r2) of 0.728 across the two techniques. Gene Ontology analyses of the differentially expressed proteins revealed that annotations related to lipid metabolic process and oxidation reduction process are overly represented in the set of under-expressed proteins in IR subjects. Furthermore, both methods quantified proteins of relevance to IR. These data also showed that SWATH-MS is a promising and compelling alternative to iTRAQ for protein quantitation of complex mixtures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cerium(IV)-Oxid-Samarium-dotiert, <0.5 μm particle size, powder, contains 20 mol % samarium as dopant
Sigma-Aldrich
Tetraethylammoniumborhydrid, technical, ≥95% (T)