Merck
  • The members of M20D peptidase subfamily from Burkholderia cepacia, Deinococcus radiodurans and Staphylococcus aureus (HmrA) are carboxydipeptidases, primarily specific for Met-X dipeptides.
The members of M20D peptidase subfamily from Burkholderia cepacia, Deinococcus radiodurans and Staphylococcus aureus (HmrA) are carboxydipeptidases, primarily specific for Met-X dipeptides.
Archives of biochemistry and biophysics (2015-10-13)
Sahayog N Jamdar, Venkata N Are, Mallikarjunan Navamani, Saurabh Kumar, Vandan Nagar, Ravindra D Makde
ZUSAMMENFASSUNG
Three members of peptidase family M20D from Burkholderia cepacia (BcepM20D; Uniprot accession no. A0A0F7GQ23), Deinococcus radiodurans R1 (DradM20D; Uniprot accession no. Q9RTP6) and Staphylococcus aureus (HmrA; Uniprot accession no. Q99Q45) were characterized in terms of their preference for various substrates. The results thus reveal that all the enzymes including HmrA lack endopeptidase as well as aminopeptidase activities and possess strong carboxypeptidase activity. Further, the amidohydrolase activity exerted on other substrates like N-Acetyl-Amino acids, N-Carbobenzoxyl-Amino acids and Indole acetic acid (IAA)-Amino acids is due to the ability of these enzymes to accommodate different types of chemical groups other than the amino acid at the S1 pocket. Further, data on peptide hydrolysis strongly suggests that all the three enzymes are primarily carboxydipeptidases exhibiting highest catalytic efficiency (kcat/Km 5-36 × 10(5) M(-1) s(-1)) for Met-X substrates, where -X could be Ala/Gly/Ser/Tyr/Phe/Leu depending on the source organism. The supportive evidence for the substrate specificities was also provided with the molecular docking studies carried out using structure of SACOL0085 and homology modelled structure of BcepM20D. The preference for different substrates, their binding at active site of the enzyme and possible role of these enzymes in recycling of methionine are discussed in this study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Imidazol, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazol, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri