Merck
Low-calorie sweeteners in food and food supplements on the Italian market.
Food additives & contaminants. Part B, Surveillance (2015-09-26)
Steven Janvier, Séverine Goscinny, Cinzia Le Donne, Joris Van Loco
ZUSAMMENFASSUNG
This study determines the occurrence and concentration levels of artificial low-calorie sweeteners (LCSs) in food and food supplements on the Italian market. The analysed sample set (290 samples) was representative of the Italian market and comprised of beverages, jams, ketchups, confectionery, dairy products, table-top sweeteners and food supplements. All samples were analysed via UPLC-MS/MS. The method was in-house validated for the analysis of seven LCSs (aspartame, acesulfame-K, saccharin, sucralose, cyclamate, neotame and neohesperidin dihydrochalcone) in food and for five LCSs (aspartame, acesulfame-K, saccharin, cyclamate and sucralose) in food supplements. Except for cyclamate in one beverage which exceeded the maximum level (ML) with 13%, all concentrations measured in food were around or below the ML. In food supplements, 40 of the 52 samples (77%) were found to be above the ML, with exceedances of up to 200% of the ML.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Supelco
Aspartam, analytical standard
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Saccharin, ≥99%
Sigma-Aldrich
Asp-Phe-OMe, ≥98%