Merck
  • Balancing the Osteogenic and Antibacterial Properties of Titanium by Codoping of Mg and Ag: An in Vitro and in Vivo Study.
Balancing the Osteogenic and Antibacterial Properties of Titanium by Codoping of Mg and Ag: An in Vitro and in Vivo Study.
ACS applied materials & interfaces (2015-07-24)
Yaochao Zhao, Huiliang Cao, Hui Qin, Tao Cheng, Shi Qian, Mengqi Cheng, Xiaochun Peng, Jiaxin Wang, Yin Zhang, Guodong Jin, Xianlong Zhang, Xuanyong Liu, Paul K Chu
ZUSAMMENFASSUNG
To simultaneously enhance the osteogenic and antibacterial properties of titanium, we introduced magnesium (Mg), silver (Ag), or both by using the plasma immersion ion implantation (PIII) technique, producing three PIII sample groups, namely, Mg-doped titanium (Mg-PIII), Ag-doped titanium (Ag-PIII), and Mg and Ag codoped titanium (Mg/Ag-PIII). The in vitro antibacterial efficacy of Mg/Ag-PIII group was about 7-10% higher than that of Ag-PIII. In vitro and in vivo results demonstrated that osteogenic property of Mg/Ag PIII group was better than that of Ag-PIII or Mg-PIII. It was believed that the galvanic effects between Mg and Ag NPs played a key role in facilitating the release of Mg but reducing the release of silver, answering for the selective performances of the Mg/Ag-PIII group over bacterial and mammalian cells. This study demonstrated that the integration of multiple functional elements could be realized by the dual-source PIII technique, and in this case, the antibacterial properties and osteogenic property of titanium could be balanced.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Tetracyclin -hydrochlorid, ≥95% (European Pharmacopoeia HPLC assay), meets EP testing specifications
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Tetracyclin -hydrochlorid, powder
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Tetracyclin -hydrochlorid, meets USP testing specifications
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%